Merck
Mechanism and function of root circumnutation.
Proceedings of the National Academy of Sciences of the United States of America (2021-02-21)
Isaiah Taylor, Kevin Lehner, Erin McCaskey, Niba Nirmal, Yasemin Ozkan-Aydin, Mason Murray-Cooper, Rashmi Jain, Elliot W Hawkes, Pamela C Ronald, Daniel I Goldman, Philip N Benfey
ABSTRACT
Early root growth is critical for plant establishment and survival. We have identified a molecular pathway required for helical root tip movement known as circumnutation. Here, we report a multiscale investigation of the regulation and function of this phenomenon. We identify key cell signaling events comprising interaction of the ethylene, cytokinin, and auxin hormone signaling pathways. We identify the gene Oryza sativa histidine kinase-1 (HK1) as well as the auxin influx carrier gene OsAUX1 as essential regulators of this process in rice. Robophysical modeling and growth challenge experiments indicate circumnutation is critical for seedling establishment in rocky soil, consistent with the long-standing hypothesis that root circumnutation facilitates growth past obstacles. Thus, the integration of robotics, physics, and biology has elucidated the functional importance of root circumnutation and uncovered the molecular mechanisms underlying its regulation.
Sigma-Aldrich
trans-Zeatin, BioReagent, suitable for plant cell culture, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Naphthaleneacetic acid, 99%
Pricing and availability is not currently available.
Supelco
Naptalam, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Zeatin hydrochloride, suitable for plant cell culture, ≥97%
Pricing and availability is not currently available.