Merck
  • Supercritical Extraction of Red Propolis: Operational Conditions and Chemical Characterization.
Supercritical Extraction of Red Propolis: Operational Conditions and Chemical Characterization.
Molecules (Basel, Switzerland) (2020-10-24)
João Henrique de Oliveira Reis, Bruna Aparecida Souza Machado, Gabriele de Abreu Barreto, Jeancarlo Pereira Dos Anjos, Larissa Moraes Dos Santos Fonseca, Alex Alisson Bandeira Santos, Fernando Luiz Pellegrini Pessoa, Janice Izabel Druzian
ABSTRACT
The objective of this study was to determine the best operational conditions for obtaining red propolis extract with high antioxidant potential through supercritical fluid extraction (SFE) technology, using carbon dioxide (CO2) as the supercritical fluid and ethanol as the cosolvent. The following parameters were studied: overall extraction curve, S/F (mass of CO2/mass of sample), cosolvent percentage (0, 1, 2 and 4%) and global yield isotherms as a function of different pressures (250, 350 and 450 bar) and temperatures (31.7, 40 and 50 °C). Within the investigated parameters, the best conditions found were an S/F of 131 and the use of ethanol at the highest concentration (4% w/w), which resulted in higher extract yields and higher content of antioxidant compounds. Formononetin, the main biomarker of red propolis, was the compound found at the highest amounts in the extracts. As expected, the temperature and pressure conditions also influenced the process yield, with 350 bar and 40 °C being the best conditions for obtaining bioactive compounds from a sample of red propolis. The novel results for red propolis found in this study show that it is possible to obtain extracts with high antioxidant potential using a clean technology under the defined conditions.
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Pricing and availability is not currently available.