Merck
  • Evaluation of microbial contamination of different pork carcass areas through culture-dependent and independent methods in small-scale slaughterhouses.
Evaluation of microbial contamination of different pork carcass areas through culture-dependent and independent methods in small-scale slaughterhouses.
International journal of food microbiology (2020-10-23)
Maria Francesca Peruzy, Kurt Houf, Marie Joossens, Zhongjia Yu, Yolande Therese Rose Proroga, Nicoletta Murru
ABSTRACT
Routine evaluation of the slaughter process is performed by the enumeration of the aerobic colony count, Enterobacteriaceae and Salmonella spp. on the carcass through destructive or non-destructive methods. With non-destructive methods, bacteria are counted from a minimum area of 100 cm2 in different sampling sites on the pork carcasses, and the results of these investigated areas are pooled to one value for the complete carcass evaluation (a total of 400 cm2). However, the composition of the bacterial community present on the different sampling areas remains unknown. The aim of the study was to characterize the microbial population present on four areas (ham, back, jowl and belly) of eight pork carcasses belonging to two different slaughterhouses through culture-dependent (Matrix-assisted laser desorption/ionization time-of-flight Mass Spectrometry MALDI-TOF MS, combined with 16S rRNA gene sequencing) and complementary culture-independent (16S rRNA amplicon sequencing) methods. The presence of Salmonella spp. and Y. enterocolitica was additionally assessed. Using MALDI-TOF MS, Staphylococcus, Pseudomonas, and Escherichia coli were found to dominate the bacterial cultures isolated from the 8 carcasses. Based on the 16S rRNA amplicon sequencing analyses however, no specific genus clearly dominated the bacterial community composition. By using this culture-independent method, the most abundant genera in microbial populations of the ham, back, jowl and belly were found to be similar, but important differences between the two slaughterhouses were observed. Thus, present data suggests that the indigenous bacterial population of individual animals is overruled by the microbial population of the slaughterhouse in which the carcass is handled. Also, our data suggests that sampling of only one carcass area by official authorities may be appropriate for the evaluation of the hygienic status of the carcasses and therefore of the slaughter process.
Product Description
Whirl-Pak® Speci-Sponge® Environmental Surface Sampling Bag, 532 mL(18 oz)
Pricing and availability is not currently available.
Millipore
Peptone Sorbitol Bile Broth, suitable for microbiology, NutriSelect® Plus
Pricing and availability is not currently available.