Merck
  • Repurposing the orphan drug nitisinone to control the transmission of African trypanosomiasis.
Repurposing the orphan drug nitisinone to control the transmission of African trypanosomiasis.
PLoS biology (2021-01-27)
Marcos Sterkel, Lee R Haines, Aitor Casas-Sánchez, Vincent Owino Adung'a, Raquel J Vionette-Amaral, Shannon Quek, Clair Rose, Mariana Silva Dos Santos, Natalia García Escude, Hanafy M Ismail, Mark I Paine, Seth M Barribeau, Simon Wagstaff, James I MacRae, Daniel Masiga, Laith Yakob, Pedro L Oliveira, Álvaro Acosta-Serrano
ABSTRACT
Tsetse transmit African trypanosomiasis, which is a disease fatal to both humans and animals. A vaccine to protect against this disease does not exist so transmission control relies on eliminating tsetse populations. Although neurotoxic insecticides are the gold standard for insect control, they negatively impact the environment and reduce populations of insect pollinator species. Here we present a promising, environment-friendly alternative to current insecticides that targets the insect tyrosine metabolism pathway. A bloodmeal contains high levels of tyrosine, which is toxic to haematophagous insects if it is not degraded and eliminated. RNA interference (RNAi) of either the first two enzymes in the tyrosine degradation pathway (tyrosine aminotransferase (TAT) and 4-hydroxyphenylpyruvate dioxygenase (HPPD)) was lethal to tsetse. Furthermore, nitisinone (NTBC), an FDA-approved tyrosine catabolism inhibitor, killed tsetse regardless if the drug was orally or topically applied. However, oral administration of NTBC to bumblebees did not affect their survival. Using a novel mathematical model, we show that NTBC could reduce the transmission of African trypanosomiasis in sub-Saharan Africa, thus accelerating current disease elimination programmes.
Product Description
Supelco
Mesotrione, PESTANAL®, analytical standard
Pricing and availability is not currently available.