Merck
  • Apigenin Alleviates Renal Fibroblast Activation through AMPK and ERK Signaling Pathways In Vitro.
Apigenin Alleviates Renal Fibroblast Activation through AMPK and ERK Signaling Pathways In Vitro.
Current pharmaceutical biotechnology (2020-03-21)
Ningning Li, Zhan Wang, Tao Sun, Yanfei Lei, Xianghua Liu, Zhenzhen Li
ABSTRACT
Renal fibrosis is a common pathway leading to the progression of chronic kidney disease. Activated fibroblasts contribute remarkably to the development of renal fibrosis. Although apigenin has been demonstrated to play a protective role from fibrotic diseases, its pharmacological effect on renal fibroblast activation remains largely unknown. Here, we examined the functional role of apigenin in the activation of renal fibroblasts response to transforming growth factor (TGF)-β1 and its potential mechanisms. Cultured renal fibroblasts (NRK-49F) were exposed to apigenin (1, 5, 10 and 20 μM), followed by the stimulation of TGF-β1 (2 ng/mL) for 24 h. The markers of fibroblast activation were determined. In order to confirm the anti-fibrosis effect of apigenin, the expression of fibrosis-associated genes in renal fibroblasts was assessed. As a consequence, apigenin alleviated fibroblast proliferation and fibroblastmyofibroblast differentiation induced by TGF-β1. Notably, apigenin significantly inhibited the fibrosis-associated genes expression in renal fibroblasts. Moreover, apigenin treatment significantly increased the phosphorylation of AMP-activated protein kinase (AMPK). Apigenin treatment also obviously reduced TGF-β1 induced phosphorylation of ERK1/2 but not Smad2/3, p38 and JNK MAPK in renal fibroblasts. In a summary, these results indicate that apigenin inhibits renal fibroblast proliferation, differentiation and function by AMPK activation and reduced ERK1/2 phosphorylation, suggesting it could be an attractive therapeutic potential for the treatment of renal fibrosis.
Sigma-Aldrich
Compound C108, ≥98% (HPLC)
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Supelco
Apigenin, analytical standard
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder