Merck
  • Phenylalanine and Tyrosine as Feed Additives for Reducing Stress and Enhancing Welfare in Gilthead Seabream and Meagre.
Phenylalanine and Tyrosine as Feed Additives for Reducing Stress and Enhancing Welfare in Gilthead Seabream and Meagre.
Animals : an open access journal from MDPI (2021-01-02)
Natalia Salamanca, Inmaculada Giráldez, Emilio Morales, Ignacio de La Rosa, Marcelino Herrera
ABSTRACT
Increased aquaculture production is associated with a growing interest in improving fish welfare. For this reason, the search for strategies to mitigate stress has intensified, one of these strategies being food supplementation with amino acids. The objective of this study was to evaluate the effects of dietary phenylalanine (Phe) and Tyrosine (Tyr) on the stress response and metabolism of juvenile gilthead seabreams (Sparus aurata) and meagres (Argyrosomus regius). Fish batches were fed a control diet and two diets supplemented with 5% Phe or Tyr for seven days. At the end of the experiment fish were stressed by air exposure for 3 min and then sacrificed for the extraction of blood and brain. Classical plasma stress markers were analyzed (glucose, lactate, proteins, cortisol), as well as hormones derived from those amino acids (adrenaline, noradrenaline and dopamine). Despite interspecific differences, fish fed the diets supplemented with Phe or Tyr showed a reduction on several stress markers. However, interspecific differences were detected for many indicators. Concretely, hormonal stress markers were significantly attenuated in meagres fed the enriched diets. Moreover, the stress condition favored a mobilization of amino acids towards the brain, especially in supplemented diets, hence this amino acid excess could be used as an energy substrate to cope with stress.