Merck
  • Thermally Switchable Electrically Conductive Thermoset rGO/PK Self-Healing Composites.
Thermally Switchable Electrically Conductive Thermoset rGO/PK Self-Healing Composites.
Polymers (2021-01-27)
Esteban Araya-Hermosilla, Alice Giannetti, Guilherme Macedo R Lima, Felipe Orozco, Francesco Picchioni, Virgilio Mattoli, Ranjita K Bose, Andrea Pucci
ABSTRACT
Among smart materials, self-healing is one of the most studied properties. A self-healing polymer can repair the cracks that occurred in the structure of the material. Polyketones, which are high-performance thermoplastic polymers, are a suitable material for a self-healing mechanism: a furanic pendant moiety can be introduced into the backbone and used as a diene for a temperature reversible Diels-Alder reaction with bismaleimide. The Diels-Alder adduct is formed at around 50 °C and broken at about 120 °C, giving an intrinsic, stimuli-responsive self-healing material triggered by temperature variations. Also, reduced graphene oxide (rGO) is added to the polymer matrix (1.6-7 wt%), giving a reversible OFF-ON electrically conductive polymer network. Remarkably, the electrical conductivity is activated when reaching temperatures higher than 100 °C, thus suggesting applications as electronic switches based on self-healing soft devices.
Sigma-Aldrich
Furfurylamine, ≥99%