Merck
  • Plasma and Urinary (Poly)phenolic Profiles after 4-Week Red Raspberry (Rubus idaeus L.) Intake with or without Fructo-Oligosaccharide Supplementation.
Plasma and Urinary (Poly)phenolic Profiles after 4-Week Red Raspberry (Rubus idaeus L.) Intake with or without Fructo-Oligosaccharide Supplementation.
Molecules (Basel, Switzerland) (2020-10-22)
Xuhuiqun Zhang, Amandeep Sandhu, Indika Edirisinghe, Britt M Burton-Freeman
ABSTRACT
Red raspberries (RRB) are high in anthocyanin- and ellagitannin- type (poly)phenols. This study aimed to investigate the effect of 4-week RRB supplementation on (poly)phenolic metabolism in adults with prediabetes and insulin-resistance (PreDM-IR); and whether adding fructo-oligosaccharides (FOS), prebiotics, would augment the microbial metabolites of RRB (poly)phenols. In a randomized crossover clinical trial, subjects (n = 35: PreDM-IR, n = 25; healthy Reference group, n = 10) consumed 1 cup RRB (fresh weight equivalence) per day and RRB with 8 g FOS per day each for 4 weeks in random order separated by 4-week washout. Plasma and urinary (poly)phenolic metabolites were characterized after (0-24h) consuming a RRB-based test drink (2 cups RRB) at baseline/week 0 and again after 4-week supplementations. A total of 123 (poly)phenolic metabolites were quantified. After 4-week RRB supplementation, several metabolite groups were significantly increased (p < 0.05), including urolithins, phenyl-γ-valerolactones, and phenolic acids. Supplementing FOS with RRB for 4 weeks enhanced benzoic acid derivatives compared to the baseline (p < 0.05). Specific effects of supplementation by metabolic status indicated 4-week RRB supplementation significantly increased microbial metabolites that were lower in PreDM-IR group. Our results suggest alterations in the capacity of PreDM-IR group to metabolize and render bioavailable raspberry-derived (poly)phenols when consumed regularly.
Sigma-Aldrich
3-(4-Hydroxy-3-methoxyphenyl)propionic acid, ≥96.0% (T)
Pricing and availability is not currently available.