Merck
IFNAR1 signaling in NK cells promotes persistent virus infection.
Science advances (2021-03-28)
Zhe Huang, Seung Goo Kang, Yunqiao Li, Jaroslav Zak, Namir Shaabani, Kaiyuan Deng, Jovan Shepherd, Raag Bhargava, John R Teijaro, Changchun Xiao
ABSTRACT
Inhibition of type 1 interferon (IFN-I) signaling promotes the control of persistent virus infection, but the underlying mechanisms remain poorly understood. Here, we report that genetic ablation of Ifnar1 specifically in natural killer (NK) cells led to elevated numbers of T follicular helper cells, germinal center B cells, and plasma cells and improved antiviral T cell function, resulting in hastened virus clearance that was comparable to IFNAR1 neutralizing antibody treatment. Antigen-specific B cells and antiviral antibodies were essential for the accelerated control of LCMV Cl13 infection following IFNAR1 blockade. IFNAR1 signaling in NK cells promoted NK cell function and general killing of antigen-specific CD4 and CD8 T cells. Therefore, inhibition of IFN-I signaling in NK cells enhances CD4 and CD8 T cell responses, promotes humoral immune responses, and thereby facilitates the control of persistent virus infection.
Sigma-Aldrich
phenylphosphoric acid, AldrichCPR
Pricing and availability is not currently available.