Merck
  • Study on corrosion behavior of X80 steel under stripping coating by sulfate reducing bacteria.
Study on corrosion behavior of X80 steel under stripping coating by sulfate reducing bacteria.
BMC biotechnology (2021-01-11)
Yan-Yu Cui, Yong-Xiang Qin, Qing-Miao Ding, Yu-Ning Gao
ABSTRACT
At present, microorganism has been considered as important factors that threaten to buried pipelines with disbonded coatings. Aiming at the problem of unknown corrosion mechanism of sulfate-reducing bacteria (SRB), a series of studies have been carried out in this paper. Spectrophotometer and fluorescent labeling technology are used to study the growth and attachment of SRB in the simulated soil solution. The corrosion behavior of X80 pipeline steel with or without SRB was researched by electrochemical methods such as open circuit potential, dynamic potential polarization curve, and electrochemical impedance spectroscopy. The microscopic morphology of the corrosion products on the surface was observed with a scanning electron microscope (SEM), and the element content of the corrosion products on the surface of the sample after corrosion was observed using X-ray energy spectrum (EDS) analysis. The results showed that the growth and reproduction of SRB caused the pH of the soil simulated solution to increase, which may promote the corrosion of X80 steel. In addition, the cathode reaction of X80 steel in a sterile environment is the reduction of H+, and the main corrosion product is iron oxide. When the soil simulation solution contains SRB, the cathodic reaction is controlled by both H+ reduction and sulfide depolarization reactions, and FeS appears in the corrosion products. Although the life cycle of SRB is only about 14 days, the corrosion of X80 steel is greatly promoted by SRB, and even causes corrosion perforation, which will bring huge economic losses and serious safety hazards.
Sigma-Aldrich
X80, ≥98% (HPLC)
Pricing and availability is not currently available.