Merck
  • Analogs of zanamivir with modified C4-substituents as the inhibitors against the group-1 neuraminidases of influenza viruses.
Analogs of zanamivir with modified C4-substituents as the inhibitors against the group-1 neuraminidases of influenza viruses.
Bioorganic & medicinal chemistry (2010-05-11)
Wen-Hsien Wen, Shi-Yun Wang, Keng-Chang Tsai, Yih-Shyun E Cheng, An-Suei Yang, Jim-Min Fang, Chi-Huey Wong
ABSTRACT
Unlike the group-2 neuraminidase, the group-1 neuraminidase of influenza virus possesses a flexible loop (the 150-loop) and a cavity (the 150-cavity) adjacent to the active site, and renders a conformational change from the 'open' form to the 'closed' form on binding with substrate (sialo-glycoprotein) or inhibitor (e.g., zanamivir). Zanamivir derivative 8a having an extended (piperazinocarbonyl)propyl substituent at the internal N-position of the guanidino group is designed as a possible inhibitor on the basis of computer docking to the open form of N1 subtype neuraminidase. Indeed, compound 8a exhibits strong neuraminidase inhibition and good anti-influenza activity against H1N1 virus with IC(50)=2.15microM and EC(50)=0.77microM, respectively. This study may provide a clue to future design of better group-1 neuraminidase inhibitors.
Sigma-Aldrich
N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid, ≥93% (TLC)
Pricing and availability is not currently available.