Merck
  • The association of urinary metabolites of polycyclic aromatic hydrocarbons with obstructive coronary artery disease: A red alert for action.
The association of urinary metabolites of polycyclic aromatic hydrocarbons with obstructive coronary artery disease: A red alert for action.
Environmental pollution (Barking, Essex : 1987) (2020-11-13)
Salwa Hajir, Lamis Al Aaraj, Nathalie Zgheib, Kamal Badr, Hussain Ismaeel, Antoine Abchee, Hani Tamim, Najat A Saliba
ABSTRACT
In Lebanon, previous studies have indicated an onset of cardiovascular diseases 12 years earlier than in other parts of the world, suggesting the presence of additional risk factors specific to Lebanon. Measurements of airborne particles in Lebanon surpass the recommendations of the World Health Organization by over 150%. This study examined the association between obstructive coronary artery disease (CAD), assessed by a novel marker calculated from coronary catheterization, and markers of air pollution, specifically polycyclic aromatic hydrocarbons (PAHs), in a cohort of 258 patients seen at the American University of Beirut Medical Center since 2014. The concentrations of four types of hydroxylated polycyclic aromatic hydrocarbons (OHPAHs), 2-OHNAP, 2-OHFLU, 3-OHPHE, and 1-OHPYR, were measured in the urine samples of these patients using high performance liquid chromatography coupled with fluorescence detector. Results showed that the OHPAH concentrations were higher than what was reported in high-income countries and, most notably, the levels for non-smokers in this study were higher than those of smokers and some occupational workers in other countries. This implies that patients were exposed to high levels of PAHs, which originate from combustion sources. In particular, 1-OHPYR showed a significant association with presence of obstructive CAD, even after adjusting for covariates like age, sex, and diabetes. Smokers or not, this association has implications for public health and calls for urgent need to pass regulations to reduce the emissions of PAH sources, such as cars, diesel generators, and incinerators.
Sigma-Aldrich
2-Hydroxyfluorene, 98%
Pricing and availability is not currently available.