Merck
  • Coaxial nanofibers of nickel/gadolinium oxide/nickel oxide as highly effective electrocatalysts for hydrogen evolution reaction.
Coaxial nanofibers of nickel/gadolinium oxide/nickel oxide as highly effective electrocatalysts for hydrogen evolution reaction.
Journal of colloid and interface science (2021-01-01)
Heba H El-Maghrabi, Amr A Nada, Maged F Bekheet, Stéphanie Roualdes, Wiebke Riedel, Igor Iatsunskyi, Emerson Coy, Aleksander Gurlo, Mikhael Bechelany
ABSTRACT
Cost-effective, active and stable electrocatalysts are crucial for hydrogen production via electrocatalytic water splitting. Here, we describe the preparation of novel nanofibers (NF) made of Ni/Gd2O3/NiO heterostructures by electrospinning. The fabricated materials showed high electrocatalytic performance for hydrogen evolution reaction (HER) with onset potential values of 89 mV, which are very close to those of platinum (Pt). NiO chemical and electronic properties were successfully optimized in Ni/Gd2O3/NiO coaxial heterostructures; NiO NFs doped with Gd3+ significantly enhanced its electrical conductivity and promoted HER reaction kinetics. These NFs offer the distinct advantages of long-term durability and readiness for hydrogen production via HER, and also better performance than benchmark Pt catalysts. The successful fabrication of these metal oxide NFs and nanostructures may represent a new approach for the rational synthesis of efficient HER catalysts.
Sigma-Aldrich
Isopropyl alcohol, natural, ≥98%, FG
Pricing and availability is not currently available.