Merck
  • Transferrin receptor 1 plays an important role in muscle development and denervation-induced muscular atrophy.
Transferrin receptor 1 plays an important role in muscle development and denervation-induced muscular atrophy.
Neural regeneration research (2020-12-16)
Ying Li, Juan-Xian Cheng, Hai-Hong Yang, Li-Ping Chen, Feng-Jiao Liu, Yan Wu, Ming Fan, Hai-Tao Wu
ABSTRACT
Previous studies demonstrate an accumulation of transferrin and transferrin receptor 1 (TfR1) in regenerating peripheral nerves. However, the expression and function of transferrin and TfR1 in the denervated skeletal muscle remain poorly understood. In this study, a mouse model of denervation was produced by complete tear of the left brachial plexus nerve. RNA-sequencing revealed that transferrin expression in the denervated skeletal muscle was upregulated, while TfR1 expression was downregulated. We also investigated the function of TfR1 during development and in adult skeletal muscles in mice with inducible deletion or loss of TfR1. The ablation of TfR1 in skeletal muscle in early development caused severe muscular atrophy and early death. In comparison, deletion of TfR1 in adult skeletal muscles did not affect survival or glucose metabolism, but caused skeletal muscle atrophy and motor functional impairment, similar to the muscular atrophy phenotype observed after denervation. These findings suggest that TfR1 plays an important role in muscle development and denervation-induced muscular atrophy. This study was approved by the Institutional Animal Care and Use Committee of Beijing Institute of Basic Medical Sciences, China (approval No. SYXK 2017-C023) on June 1, 2018.
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Slow) antibody produced in mouse, clone NOQ7.5.4D, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Doxycycline hyclate
Pricing and availability is not currently available.
Magnetic Stir Bar, for use with For 3.0-5.0mL vessels, pkg of 6 ea, PTFE
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid
Pricing and availability is not currently available.