Merck
  • Synthesis of Conserved Odor Object Representations in a Random, Divergent-Convergent Network.
Synthesis of Conserved Odor Object Representations in a Random, Divergent-Convergent Network.
Neuron (2020-08-20)
Keita Endo, Yoshiko Tsuchimoto, Hokto Kazama
ABSTRACT
Animals are capable of recognizing mixtures and groups of odors as a unitary object. However, how odor object representations are generated in the brain remains elusive. Here, we investigate sensory transformation between the primary olfactory center and its downstream region, the mushroom body (MB), in Drosophila and show that clustered representations for mixtures and groups of odors emerge in the MB at the population and single-cell levels. Decoding analyses demonstrate that neurons selective for mixtures and groups enhance odor generalization. Responses of these neurons and those selective for individual odors all emerge in an experimentally well-constrained model implementing divergent-convergent, random connectivity between the primary center and the MB. Furthermore, we found that relative odor representations are conserved across animals despite this random connectivity. Our results show that the generation of distinct representations for individual odors and groups and mixtures of odors in the MB can be understood in a unified computational and mechanistic framework.
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%
Sigma-Aldrich
Valeric acid, ≥99%