Merck
  • A higher voltage Fe(ii) bipyridine complex for non-aqueous redox flow batteries.
A higher voltage Fe(ii) bipyridine complex for non-aqueous redox flow batteries.
Dalton transactions (Cambridge, England : 2003) (2020-12-22)
Claudina X Cammack, Harry D Pratt, Leo J Small, Travis M Anderson
ABSTRACT
Non-aqueous redox flow batteries (RFBs) offer the possibility of higher voltage and a wider working temperature range than their aqueous counterpart. Here, we optimize the established 2.26 V Fe(bpy)3(BF4)2/Ni(bpy)3(BF4)2 asymmetric RFB to lessen capacity fade and improve energy efficiency over 20 cycles. We also prepared a family of substituted Fe(bpyR)3(BF4)2 complexes (R = -CF3, -CO2Me, -Br, -H, -tBu, -Me, -OMe, -NH2) to potentially achieve a higher voltage RFB by systematically tuning the redox potential of Fe(bpyR)3(BF4)2, from 0.94 V vs. Ag/AgCl for R = OMe to 1.65 V vs. Ag/AgCl for R = CF3 (ΔV = 0.7 V). A series of electronically diverse symmetric and asymmetric RFBs were compared and contrasted to study electroactive species stability and efficiency, in which the unsubstituted Fe(bpy)3(BF4)2 exhibited the highest stability as a catholyte in both symmetric and asymmetric cells with voltage and coulombic efficiencies of 94.0% and 96.5%, and 90.7% and 80.7%, respectively.
Sigma-Aldrich
Tetrabutylammonium trifluoromethanesulfonate, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
4,4′-Di-tert-butyl-2,2′-dipyridyl, 98%
Pricing and availability is not currently available.