Merck
  • Mechanochromic and Thermochromic Sensors Based on Graphene Infused Polymer Opals.
Mechanochromic and Thermochromic Sensors Based on Graphene Infused Polymer Opals.
Advanced functional materials (2020-08-11)
Izabela Jurewicz, Alice A K King, Ravi Shanker, Matthew J Large, Ronan J Smith, Ross Maspero, Sean P Ogilvie, Jurgen Scheerder, Jun Han, Claudia Backes, Joselito M Razal, Marian Florescu, Joseph L Keddie, Jonathan N Coleman, Alan B Dalton
ABSTRACT
High quality opal-like photonic crystals containing graphene are fabricated using evaporation-driven self-assembly of soft polymer colloids. A miniscule amount of pristine graphene within a colloidal crystal lattice results in the formation of colloidal crystals with a strong angle-dependent structural color and a stop band that can be reversibly shifted across the visible spectrum. The crystals can be mechanically deformed or can reversibly change color as a function of their temperature, hence their sensitive mechanochromic and thermochromic response make them attractive candidates for a wide range of visual sensing applications. In particular, it is shown that the crystals are excellent candidates for visual strain sensors or integrated time-temperature indicators which act over large temperature windows. Given the versatility of these crystals, this method represents a simple, inexpensive, and scalable approach to produce multifunctional graphene infused synthetic opals and opens up exciting applications for novel solution-processable nanomaterial based photonics.