Merck
  • Cloning, characterization, and inhibition studies of a beta-carbonic anhydrase from Brucella suis.
Cloning, characterization, and inhibition studies of a beta-carbonic anhydrase from Brucella suis.
Journal of medicinal chemistry (2010-02-18)
Pascale Joseph, François Turtaut, Safia Ouahrani-Bettache, Jean-Louis Montero, Isao Nishimori, Tomoko Minakuchi, Daniela Vullo, Andrea Scozzafava, Stephan Köhler, Jean-Yves Winum, Claudiu T Supuran
ABSTRACT
A beta-carbonic anhydrase (CA, EC 4.2.1.1) from the bacterial pathogen Brucella suis, bsCA 1, has been cloned, purified, and characterized kinetically. bsCA 1 has appreciable activity as catalyst for the hydration of CO(2) to bicarbonate, with a k(cat) of 6.4 x 10(5) s(-1) and k(cat)/K(m) of 3.9 x 10(7) M(-1).s(-1). A panel of 38 sulfonamides and one sulfamate have been investigated for inhibition of this new beta-CA. All types of activities have been detected, with K(I)s in the range of 17 nM to 5.87 microM. The best bsCA 1 inhibitors were ethoxzolamide (17 nM), celecoxib (18 nM), dorzolamide (21 nM), valdecoxib, and sulpiride (19 nM). Whether bsCA 1 inhibitors may have application in the fight against brucellosis, an endemic disease and the major bacterial zoonosis, producing debilitating infection in humans and animals, warrants further studies.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
4-Sulfamoylbenzoic acid, 97%
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%