Merck
  • Human pharmaceutical and pesticide residues in Israeli dairy milk in association with dietary risk assessment.
Human pharmaceutical and pesticide residues in Israeli dairy milk in association with dietary risk assessment.
Food additives & contaminants. Part B, Surveillance (2020-05-15)
Vijayakumar Bommuraj, Yaira Chen, Orit Gal, Julius Ben Ari, Evgenia Kertsnus-Banchik, Shimon Barel, Jakob A Shimshoni
ABSTRACT
Throughout the world, more than six billion people consume milk and milk products yearly. The safety and quality of dairy milk are regularly monitored in most countries worldwide. The Israeli monitoring program of chemical residues in milk has not changed in the last decades, focusing only on major veterinary drugs and few selected environmental contaminants such as heavy metals and persistent organic pollutants. Consequently, a knowledge gap exists regarding the potential occurrence of other chemicals such as human pharmaceuticals and non-monitored pesticides in milk. In this survey, 51 commercial bovine and goat milk samples were analysed by LC-MS/MS and pharmaceutical and pesticide residues are reported in the range of 0.1-93 µg/L. Israeli milk samples revealed at least one and up to five chemical residues simultaneously. The pesticides found in milk were below the European maximum residue limit values. The risk assessment performed, indicated negligible risk.
Product Description
Supelco
Imidacloprid-d4, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Acetamiprid-d3, PESTANAL®, analytical standard
Pricing and availability is not currently available.