Merck
  • Central angiotensin-(1-7) attenuates systemic inflammation via activation of sympathetic signaling in endotoxemic rats.
Central angiotensin-(1-7) attenuates systemic inflammation via activation of sympathetic signaling in endotoxemic rats.
Brain, behavior, and immunity (2020-04-27)
Patrícia Passaglia, Felipe de Lima Faim, Marcelo Eduardo Batalhão, Lusiane Maria Bendhack, José Antunes-Rodrigues, Luis Ulloa, Alexandre Kanashiro, Evelin Capellari Carnio
ABSTRACT
Angiotensin-(1-7) [Ang-(1-7)] is an angiotensin-derived neuropeptide with potential anti-hypertensive and anti-inflammatory properties. However, a possible action of Ang-(1-7) in neuroimmune interactions to regulate inflammatory response has not been explored. Thus, the aim of this study was to determine whether the intracerebroventricular (i.c.v.) administration of Ang-(1-7) can modulate systemic inflammation via sympathetic efferent circuits. Wistar male rats received systemic administration of lipopolysaccharide (LPS) (1.5 mg/Kg). Ang-(1-7) (0.3 nmol in 2 µL) promoted the release of splenic norepinephrine and attenuated tumor necrosis factor (TNF) and nitric oxide (NO), but increased interleukin-10 (IL-10), levels in the serum, spleen, and liver in endotoxemic rats. Furthermore, 6-hydroxydopamine-induced chemical sympathectomy (100 mg/Kg, intravenous) or i.c.v. administration of Mas receptor antagonist A779 (3 nmol in 2 µL) abolished the anti-inflammatory effects of central Ang-(1-7) injection. Moreover, this treatment did not alter the plasmatic LPS-induced corticosterone and vasopressin. The administration of Ang-(1-7) reverted the low resistance in response to catecholamines of rings of thoracic aorta isolated from endotoxemic rats, treated or not, with this peptide by a mechanism dependent on the regulation of NO released from perivascular adipose tissue. Together, our results indicate that Ang-(1-7) regulates systemic inflammation and vascular hyporesponsiveness in endotoxemia via activation of a central Mas receptors/sympathetic circuits/norepinephrine axis and provide novel mechanistic insights into the anti-inflammatory Ang-(1-7) properties.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Angiotensin Fragment 1-7 acetate salt hydrate, ≥90% (HPLC), powder