Merck
  • Establishment and maintenance of motor neuron identity via temporal modularity in terminal selector function.
Establishment and maintenance of motor neuron identity via temporal modularity in terminal selector function.
eLife (2020-10-02)
Yinan Li, Anthony Osuma, Edgar Correa, Munachiso A Okebalama, Pauline Dao, Olivia Gaylord, Jihad Aburas, Priota Islam, André Ex Brown, Paschalis Kratsios
ABSTRACT
Terminal selectors are transcription factors (TFs) that establish during development and maintain throughout life post-mitotic neuronal identity. We previously showed that UNC-3/Ebf, the terminal selector of C. elegans cholinergic motor neurons (MNs), acts indirectly to prevent alternative neuronal identities (Feng et al., 2020). Here, we globally identify the direct targets of UNC-3. Unexpectedly, we find that the suite of UNC-3 targets in MNs is modified across different life stages, revealing 'temporal modularity' in terminal selector function. In all larval and adult stages examined, UNC-3 is required for continuous expression of various protein classes (e.g. receptors, transporters) critical for MN function. However, only in late larvae and adults, UNC-3 is required to maintain expression of MN-specific TFs. Minimal disruption of UNC-3's temporal modularity via genome engineering affects locomotion. Another C. elegans terminal selector (UNC-30/Pitx) also exhibits temporal modularity, supporting the potential generality of this mechanism for the control of neuronal identity.
Product Description
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Pricing and availability is not currently available.