Merck
  • Wnt3 distribution in the zebrafish brain is determined by expression, diffusion and multiple molecular interactions.
Wnt3 distribution in the zebrafish brain is determined by expression, diffusion and multiple molecular interactions.
eLife (2020-11-26)
Sapthaswaran Veerapathiran, Cathleen Teh, Shiwen Zhu, Indira Kartigayen, Vladimir Korzh, Paul T Matsudaira, Thorsten Wohland
ABSTRACT
Wnt3 proteins are lipidated and glycosylated signaling molecules that play an important role in zebrafish neural patterning and brain development. However, the transport mechanism of lipid-modified Wnts through the hydrophilic extracellular environment for long-range action remains unresolved. Here we determine how Wnt3 accomplishes long-range distribution in the zebrafish brain. First, we characterize the Wnt3-producing source and Wnt3-receiving target regions. Subsequently, we analyze Wnt3 mobility at different length scales by fluorescence correlation spectroscopy and fluorescence recovery after photobleaching. We demonstrate that Wnt3 spreads extracellularly and interacts with heparan sulfate proteoglycans (HSPG). We then determine the binding affinity of Wnt3 to its receptor, Frizzled1 (Fzd1), using fluorescence cross-correlation spectroscopy and show that the co-receptor, low-density lipoprotein receptor-related protein 5 (Lrp5), is required for Wnt3-Fzd1 interaction. Our results are consistent with the extracellular distribution of Wnt3 by a diffusive mechanism that is modified by tissue morphology, interactions with HSPG, and Lrp5-mediated receptor binding, to regulate zebrafish brain development.
Sigma-Aldrich
Surfen hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Heparinase I from Flavobacterium heparinum, Lyophilized powder stabilized with approx. 25% bovine serum albumin, ≥200 units/mg protein (enzyme + BSA)
Pricing and availability is not currently available.