Merck
  • Runx2+ Niche Cells Maintain Incisor Mesenchymal Tissue Homeostasis through IGF Signaling.
Runx2+ Niche Cells Maintain Incisor Mesenchymal Tissue Homeostasis through IGF Signaling.
Cell reports (2020-08-14)
Shuo Chen, Junjun Jing, Yuan Yuan, Jifan Feng, Xia Han, Quan Wen, Thach-Vu Ho, Chelsea Lee, Yang Chai
ABSTRACT
Stem cell niches provide a microenvironment to support the self-renewal and multi-lineage differentiation of stem cells. Cell-cell interactions within the niche are essential for maintaining tissue homeostasis. However, the niche cells supporting mesenchymal stem cells (MSCs) are largely unknown. Using single-cell RNA sequencing, we show heterogeneity among Gli1+ MSCs and identify a subpopulation of Runx2+/Gli1+ cells in the adult mouse incisor. These Runx2+/Gli1+ cells are strategically located between MSCs and transit-amplifying cells (TACs). They are not stem cells but help to maintain the MSC niche via IGF signaling to regulate TAC proliferation, differentiation, and incisor growth rate. ATAC-seq and chromatin immunoprecipitation reveal that Runx2 directly binds to Igfbp3 in niche cells. This Runx2-mediated IGF signaling is crucial for regulating the MSC niche and maintaining tissue homeostasis to support continuous growth of the adult mouse incisor, providing a model for analysis of the molecular regulation of the MSC niche.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Insulin-like Growth Factor-II from mouse, IGF-II, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
Toluidine Blue O, Technical grade