Merck
  • Synthesis and evaluation of novel bis-pyridinium oximes as reactivators of DFP-inhibited acetylcholinesterase.
Synthesis and evaluation of novel bis-pyridinium oximes as reactivators of DFP-inhibited acetylcholinesterase.
European journal of medicinal chemistry (2008-04-09)
Jyotiranjan Acharya, Arvind Kumar Gupta, Devendra Kumar Dubey, Syed Kalbey Raza
ABSTRACT
A series of novel bis-pyridinium oximes connected by bis-methoxymethyl benzene, 1,4-bis-methoxymethyl (cis)-but-2-ene and 1,4-bis-methoxymethyl but-2-yne linkers were synthesized and their in vitro reactivation efficacy was evaluated against diisopropyl phosphorofluoridate (DFP) inhibited acetylcholinesterase (AChE) and compared with the established antidote 2-PAM and obidoxime. However, the best reactivation was observed with the standard oxime 2-PAM. The reactivation efficacy of 1,3-dimethoxymethyl benzene bis-[4,4'-(hydroxyiminomethyl) pyridinium] dichloride (3d) and 1,4-dimethoxy but-2-ene bis-[4,4'-(hydroxyiminomethyl) pyridinium] dichloride (3g) was comparable with that of obidoxime, another standard antidote.
Sigma-Aldrich
Pyridine-2-aldoxime methochloride
Pricing and availability is not currently available.