Merck
  • Prognostic significance of Beclin 1 in intrahepatic cholangiocellular carcinoma.
Prognostic significance of Beclin 1 in intrahepatic cholangiocellular carcinoma.
Autophagy (2011-06-10)
Li-Wei Dong, Yu-Jie Hou Hou, Ye-Xiong Tan, Liang Tang, Yu-Fei Pan, Min Wang, Hong-Yang Wang
ABSTRACT
Autophagy enables cells to recycle long-lived proteins or damaged organelles. Beclin 1 plays important roles in autophagy, differentiation, apoptosis and the development and progression of cancer, but the expression of Beclin 1 and its possible role in primary intrahepatic cholangiocarcinoma (ICC) has not been reported yet. This study aimed to investigate Beclin 1 expression and its prognostic significance in ICC. First, we assessed the expression levels of Becn1 by real-time PCR in 50 ICC samples and found Becn1 mRNA expression was markedly increased in 78% (39 of 50) samples compared with normal bile duct epithelium. Beclin 1 protein expression in 108 tumor specimens from patients diagnosed with ICC was examined by immunohistochemistry and the correlation between Beclin 1 expression and clinicopathological factors were investigated. Immunopositivity for Beclin 1 was found in 72.2% (78 of 108) samples and low Beclin 1 expression was significantly associated with lymph node metastasis. The correlation between Beclin 1 expression and metastasis was validated in 46 ICC samples with lymph node metastasis. In survival analysis, low Beclin 1 expression was associated with worse overall survival (OS; p = 0.025) and disease-free survival (DFS; p = 0.027). In multivariate analysis, Beclin 1 expression, intrahepatic metastasis, lymph node metastasis and tumor size were found to be independent prognostic factors of OS. Thus, our results suggested the expression of Beclin 1 was correlated with progression and metastasis of ICC and it might serve as a novel prognostic marker for patients with ICC.
Sigma-Aldrich
IHC Select Secondary Goat anti-Mouse IgG, anti-Rabbit IgG, biotinylated, from goat
Pricing and availability is not currently available.