Merck
  • ERK2 phosphorylates Krüppel-like factor 8 protein at serine 48 to maintain its stability.
ERK2 phosphorylates Krüppel-like factor 8 protein at serine 48 to maintain its stability.
American journal of cancer research (2016-06-14)
Satadru K Lahiri, Heng Lu, Debarati Mukherjee, Lin Yu, Jihe Zhao
ABSTRACT
Krüppel-like factor 8 (KLF8) plays important roles in cancer and is strictly regulated by various post-translational modifications such as sumoylation, acetylation, ubiquitylation and PARylation. Here we report a novel phosphorylation of KLF8 by ERK2 responsible and critical for the stability of KLF8 protein. The full-length KLF8 protein displays a doublet in SDS-PAGE gel. The upper band of the doublet, however, disappeared when the N-terminal 50 amino acids were deleted. In its full-length the upper band disappeared upon phosphatase treatment or mutation of the serine 48 (S48) to alanine whereas the lower band was lost when the S48 was mutated to aspartic acid that mimics phosphorylated S48. These results suggest that S48 phosphorylation is responsible for the motility up-shift of KLF8 protein. Pharmacological and genetic manipulations of various potential kinases identified ERK2 as the likely one that phosphorylates KLF8 at S48. Functional studies indicated that this phosphorylation is crucial for protecting KLF8 protein from degradation in the nucleus and promoting cell migration. Taken together, this study identifies a novel mechanism of phosphorylation critical for KLF8 protein stabilization and function.
Product Description
Millipore
Anti-HA Immunoprecipitation Kit
Pricing and availability is not currently available.