Merck
  • Single-cell RNA sequencing shows the immunosuppressive landscape and tumor heterogeneity of HBV-associated hepatocellular carcinoma.
Single-cell RNA sequencing shows the immunosuppressive landscape and tumor heterogeneity of HBV-associated hepatocellular carcinoma.
Nature communications (2021-06-19)
Daniel Wai-Hung Ho, Yu-Man Tsui, Lo-Kong Chan, Karen Man-Fong Sze, Xin Zhang, Jacinth Wing-Sum Cheu, Yung-Tuen Chiu, Joyce Man-Fong Lee, Albert Chi-Yan Chan, Elaine Tin-Yan Cheung, Derek Tsz-Wai Yau, Nam-Hung Chia, Irene Lai-Oi Lo, Pak-Chung Sham, Tan-To Cheung, Carmen Chak-Lui Wong, Irene Oi-Lin Ng
ABSTRACT
Interaction between tumor cells and immune cells in the tumor microenvironment is important in cancer development. Immune cells interact with the tumor cells to shape this process. Here, we use single-cell RNA sequencing analysis to delineate the immune landscape and tumor heterogeneity in a cohort of patients with HBV-associated human hepatocellular carcinoma (HCC). We found that tumor-associated macrophages suppress tumor T cell infiltration and TIGIT-NECTIN2 interaction regulates the immunosuppressive environment. The cell state transition of immune cells towards a more immunosuppressive and exhaustive status exemplifies the overall cancer-promoting immunocellular landscape. Furthermore, the heterogeneity of global molecular profiles reveals co-existence of intra-tumoral and inter-tumoral heterogeneity, but is more apparent in the latter. This analysis of the immunosuppressive landscape and intercellular interactions provides mechanistic information for the design of efficacious immune-oncology treatments in hepatocellular carcinoma.
Sigma-Aldrich
Anti-LAIR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-NECTIN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Millipore
Millicell® 24 well Plate, pore size 0.4 μm, polycarbonate membrane, sterile
Pricing and availability is not currently available.