Merck
  • Trend analysis of a database of intravenous pharmacokinetic parameters in humans for 670 drug compounds.
Trend analysis of a database of intravenous pharmacokinetic parameters in humans for 670 drug compounds.
Drug metabolism and disposition: the biological fate of chemicals (2008-04-23)
R Scott Obach, Franco Lombardo, Nigel J Waters
ABSTRACT
We present herein a compilation and trend analysis of human i.v. pharmacokinetic data on 670 drugs representing, to our knowledge, the largest publicly available set of human clinical pharmacokinetic data. This data set provides the drug metabolism scientist with a robust and accurate resource suitable for a number of applications, including in silico modeling, in vitro-in vivo scaling, and physiologically based pharmacokinetic approaches. Clearance, volume of distribution at steady state, mean residence time, and terminal phase half-life were obtained or derived from original references exclusively from studies utilizing i.v. administration. Plasma protein binding data were collected from other sources to supplement these pharmacokinetic data. These parameters were analyzed concurrently with a range of simple physicochemical descriptors, and resultant trends and patterns within the data are presented. Our findings with this much expanded data set were consistent with earlier described notions of trends between physicochemical properties and pharmacokinetic behavior. These observations and analyses, along with the large database of human pharmacokinetic data, should enable future efforts aimed toward developing quantitative structure-pharmacokinetic relationships and improving our understanding of the relationship between fundamental chemical characteristics and drug disposition.
Sigma-Aldrich
Lithium carbonate, puriss. p.a., ACS reagent, reagent (for microscopy), ≥99.0% (T)
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Supelco
Betamethasone, VETRANAL®, analytical standard
Supelco
Temozolomide, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Sulfisoxazole, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Dapsone, VETRANAL®, analytical standard
Sigma-Aldrich
Hypericin from Hypericum perforatum, ~95% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Supelco
Salbutamol, VETRANAL®, analytical standard
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
Sparfloxacin, ≥98.0% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Salbutamol