Merck
Multiple prebiotic metals mediate translation.
Proceedings of the National Academy of Sciences of the United States of America (2018-11-11)
Marcus S Bray, Timothy K Lenz, Jay William Haynes, Jessica C Bowman, Anton S Petrov, Amit R Reddi, Nicholas V Hud, Loren Dean Williams, Jennifer B Glass
ABSTRACT
Today, Mg2+ is an essential cofactor with diverse structural and functional roles in life's oldest macromolecular machine, the translation system. We tested whether ancient Earth conditions (low O2, high Fe2+, and high Mn2+) can revert the ribosome to a functional ancestral state. First, SHAPE (selective 2'-hydroxyl acylation analyzed by primer extension) was used to compare the effect of Mg2+, Fe2+, and Mn2+ on the tertiary structure of rRNA. Then, we used in vitro translation reactions to test whether Fe2+ or Mn2+ could mediate protein production, and quantified ribosomal metal content. We found that (i) Mg2+, Fe2+, and Mn2+ had strikingly similar effects on rRNA folding; (ii) Fe2+ and Mn2+ can replace Mg2+ as the dominant divalent cation during translation of mRNA to functional protein; and (iii) Fe and Mn associate extensively with the ribosome. Given that the translation system originated and matured when Fe2+ and Mn2+ were abundant, these findings suggest that Fe2+ and Mn2+ played a role in early ribosomal evolution.
Sigma-Aldrich
Dihydrofolate Reductase Assay Kit, 1 kit sufficient for 50-100 tests
Pricing and availability is not currently available.