Merck
  • Expression and Localization of Cathepsins B, D, and G in Dupuytren's Disease.
Expression and Localization of Cathepsins B, D, and G in Dupuytren's Disease.
Plastic and reconstructive surgery. Global open (2018-04-05)
Kirin Tan, Helen D Brasch, Bede van Schaijik, James R Armstrong, Reginald W Marsh, Paul F Davis, Swee T Tan, Tinte Itinteang
ABSTRACT
The pathogenesis of Dupuytren's disease (DD) remains unclear. An embryonic stem cell (ESC)-like population in the endothelium of the microvessels around tissues that expresses components of the renin-angiotensin system (RAS) has been reported. This study investigated if this primitive population expresses cathepsins B, D, and G, that contribute to RAS bypass loops. 3,3-Diaminobenzidine immunohistochemical (IHC) staining for cathepsins B, D, and G was performed on sections of formalin-fixed paraffin-embedded DD cords (n = 10) and nodules (n = 10). Immunofluorescence IHC staining was utilized to demonstrate co-expression of these cathepsins with ESC markers. Protein and gene expression of these cathepsins was investigated in snap-frozen DD cords (n = 3) and nodules (n = 3) by Western blotting and NanoString analysis, respectively. Enzymatic activity of these cathepsins was investigated by enzymatic activity assays. 3,3-Diaminobenzidine IHC staining demonstrated expression of cathepsins B, D, and G in DD cords and nodules. Gene expression of cathepsins B, D, and G was confirmed by NanoString analysis. Western blotting confirmed expression of cathepsins B and D, but not cathepsin G. Immunofluorescent IHC staining demonstrated high abundance of cathepsins B and D on the OCT4+/angiotensin converting enzyme+ endothelium and the smooth muscle layer of the microvessels. Cathepsin G was localized to trypase+ cells within the stroma in DD cords and nodules with limited expression on the microvessels. Enzyme activity assays demonstrated functional activity of cathepsins B and D. Cathepsins B, D, and G were expressed in the DD tissues, with cathepsins B and D localized to the primitive population in the endothelium of the microvessels, whereas cathepsin G was localized to phenotypic mast cells, suggesting the presence of bypass loops for the RAS.
Product Description
Millipore
ProteoExtract® Protein Precipitation Kit
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O