Merck
  • Glucagon-like Peptide 1 Receptor Signaling in Acinar Cells Causes Growth-Dependent Release of Pancreatic Enzymes.
Glucagon-like Peptide 1 Receptor Signaling in Acinar Cells Causes Growth-Dependent Release of Pancreatic Enzymes.
Cell reports (2016-12-16)
Nicolai J Wewer Albrechtsen, Reidar Albrechtsen, Lasse Bremholm, Berit Svendsen, Rune E Kuhre, Steen S Poulsen, Charlotte B Christiansen, Elisa P Jensen, Charlotte Janus, Linda Hilsted, Carolyn F Deacon, Bolette Hartmann, Jens J Holst
ABSTRACT
Incretin-based therapies are widely used for type 2 diabetes and now also for obesity, but they are associated with elevated plasma levels of pancreatic enzymes and perhaps a modestly increased risk of acute pancreatitis. However, little is known about the effects of the incretin hormone glucagon-like peptide 1 (GLP-1) on the exocrine pancreas. Here, we identify GLP-1 receptors on pancreatic acini and analyze the impact of receptor activation in humans, rodents, isolated acini, and cell lines from the exocrine pancreas. GLP-1 did not directly stimulate amylase or lipase release. However, we saw that GLP-1 induces phosphorylation of the epidermal growth factor receptor and activation of Foxo1, resulting in cell growth with concomitant enzyme release. Our work uncovers GLP-1-induced signaling pathways in the exocrine pancreas and suggests that increases in amylase and lipase levels in subjects treated with GLP-1 receptor agonists reflect adaptive growth rather than early-stage pancreatitis.
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for IM IV, sufficient for 100 assays, Kit size M
Pricing and availability is not currently available.