Merck
  • Stimulation of 3D osteogenesis by mesenchymal stem cells using a nanovibrational bioreactor.
Stimulation of 3D osteogenesis by mesenchymal stem cells using a nanovibrational bioreactor.
Nature biomedical engineering (2017-09-01)
Penelope M Tsimbouri, Peter G Childs, Gabriel D Pemberton, Jingli Yang, Vineetha Jayawarna, Wich Orapiriyakul, Karl Burgess, Cristina González-García, Gavin Blackburn, Dilip Thomas, Catalina Vallejo-Giraldo, Manus J P Biggs, Adam S G Curtis, Manuel Salmerón-Sánchez, Stuart Reid, Matthew J Dalby
ABSTRACT
Bone grafts are one of the most commonly transplanted tissues. However, autologous grafts are in short supply, and can be associated with pain and donor-site morbidity. The creation of tissue-engineered bone grafts could help to fulfil clinical demand and provide a crucial resource for drug screening. Here, we show that vibrations of nanoscale amplitude provided by a newly developed bioreactor can differentiate a potential autologous cell source, mesenchymal stem cells (MSCs), into mineralized tissue in 3D. We demonstrate that nanoscale mechanotransduction can stimulate osteogenesis independently of other environmental factors, such as matrix rigidity. We show this by generating mineralized matrix from MSCs seeded in collagen gels with stiffness an order of magnitude below the stiffness of gels needed to induce bone formation in vitro. Our approach is scalable and can be compatible with 3D scaffolds.
Product Description
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Beta Catenin Antibody, clone 7F7.2, clone 7F7.2, Upstate®, from mouse
Pricing and availability is not currently available.