Merck
  • Anti-tumor effects of an ID antagonist with no observed acquired resistance.
Anti-tumor effects of an ID antagonist with no observed acquired resistance.
NPJ breast cancer (2021-05-26)
Paulina M Wojnarowicz, Marta Garcia Escolano, Yun-Han Huang, Bina Desai, Yvette Chin, Riddhi Shah, Sijia Xu, Saurabh Yadav, Sergey Yaklichkin, Ouathek Ouerfelli, Rajesh Kumar Soni, John Philip, David C Montrose, John H Healey, Vinagolu K Rajasekhar, William A Garland, Jeremy Ratiu, Yuan Zhuang, Larry Norton, Neal Rosen, Ronald C Hendrickson, Xi Kathy Zhou, Antonio Iavarone, Joan Massague, Andrew J Dannenberg, Anna Lasorella, Robert Benezra
ABSTRACT
ID proteins are helix-loop-helix (HLH) transcriptional regulators frequently overexpressed in cancer. ID proteins inhibit basic-HLH transcription factors often blocking differentiation and sustaining proliferation. A small-molecule, AGX51, targets ID proteins for degradation and impairs ocular neovascularization in mouse models. Here we show that AGX51 treatment of cancer cell lines impairs cell growth and viability that results from an increase in reactive oxygen species (ROS) production upon ID degradation. In mouse models, AGX51 treatment suppresses breast cancer colonization in the lung, regresses the growth of paclitaxel-resistant breast tumors when combined with paclitaxel and reduces tumor burden in sporadic colorectal neoplasia. Furthermore, in cells and mice, we fail to observe acquired resistance to AGX51 likely the result of the inability to mutate the binding pocket without loss of ID function and efficient degradation of the ID proteins. Thus, AGX51 is a first-in-class compound that antagonizes ID proteins, shows strong anti-tumor effects and may be further developed for the management of multiple cancers.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
AGX51, ≥98% (HPLC)
Sigma-Aldrich
Erastin, A cell-permeable piperazinyl-quinazolinone compound that exhibits oncogene-selective lethality.
Sigma-Aldrich
Anti-α-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Necrostatin-1, Necrostatin-1, CAS 4311-88-0, is a cell-permeable, potent, and selective blocker of necroptosis (EC50 = 494 nM in FADD-deficient Jurkat cells treated with TNF-α).