Merck
  • Differential human antibody repertoires following Zika infection and the implications for serodiagnostics and disease outcome.
Differential human antibody repertoires following Zika infection and the implications for serodiagnostics and disease outcome.
Nature communications (2019-04-28)
Supriya Ravichandran, Megan Hahn, Pablo F Belaunzarán-Zamudio, José Ramos-Castañeda, Gabriel Nájera-Cancino, Sandra Caballero-Sosa, Karla R Navarro-Fuentes, Guillermo Ruiz-Palacios, Hana Golding, John H Beigel, Surender Khurana
ABSTRACT
Zika virus (ZIKV) outbreak in Americas led to extensive efforts to develop vaccines and ZIKV-specific diagnostics. In the current study, we use whole genome phage display library spanning the entire ZIKV genome (ZIKV-GFPDL) for in-depth immune profiling of IgG and IgM antibody repertoires in serum and urine longitudinal samples from individuals acutely infected with ZIKV. We observe a very diverse IgM immune repertoire encompassing the entire ZIKV polyprotein on day 0 in both serum and urine. ZIKV-specific IgG antibodies increase 10-fold between day 0 and day 7 in serum, but not in urine; these are highly focused on prM/E, NS1 and NS2B. Differential antibody affinity maturation is observed against ZIKV structural E protein compared with nonstructural protein NS1. Serum antibody affinity to ZIKV-E protein inversely correlates with ZIKV disease symptoms. Our study provides insight into unlinked evolution of immune response to ZIKV infection and identified unique targets for ZIKV serodiagnostics.
Sigma-Aldrich
Anti-Zika Virus Antibody, clone ZV-54, from mouse