Merck
  • Chronic exposure of interleukin-13 suppress the induction of matrix metalloproteinase-1 by tumour necrosis factor α in normal and scleroderma dermal fibroblasts through protein kinase B/Akt.
Chronic exposure of interleukin-13 suppress the induction of matrix metalloproteinase-1 by tumour necrosis factor α in normal and scleroderma dermal fibroblasts through protein kinase B/Akt.
Clinical and experimental immunology (2017-09-09)
M L Brown Lobbins, B R Shivakumar, A E Postlethwaite, K A Hasty
ABSTRACT
Peripheral blood mononuclear cells taken from patients with scleroderma express increased levels of interleukin (IL)-13. Moreover, the expression of matrix metalloproteinase-1 (MMP-1) from involved scleroderma skin fibroblasts is refractory to stimulation by tumour necrosis factor (TNF)-α. To elucidate the mechanism(s) involved, we examined the effect of IL-13 on TNF-α-induced MMP-1 expression in normal and scleroderma human dermal fibroblast lines and studied the involvement of serine/threonine kinase B/protein kinase B (Akt) in this response. Dermal fibroblast lines were stimulated with TNF-α in the presence of varying concentrations of IL-13. Total Akt and pAkt were quantitated using Western blot analyses. Fibroblasts were treated with or without Akt inhibitor VIII in the presence of IL-13 followed by TNF-α stimulation. MMP-1 expression was analysed by real-time polymerase chain reaction (PCR) and enzyme-linked immunosorbent assay (ELISA). Statistical analysis was performed using analysis of variance (anova) or Student's t-test. Upon TNF-α stimulation, normal dermal fibroblasts secrete more MMP-1 than systemic sclerosis (SSc) fibroblasts. This increase in MMP-1 is lost when fibroblasts are co-incubated with IL-13 and TNF-α. IL-13 induced a significant increase in levels of pAkt in dermal fibroblasts, while Akt inhibitor VIII reversed the suppressive effects of IL-13 on the response of cultured fibroblasts to TNF-α, increasing their expression of MMP-1. We show that IL-13 suppresses MMP-1 in TNF-α-stimulated normal and scleroderma dermal fibroblast. Akt inhibitor VIII is able to reverse the suppressive effect of IL-13 on MMP-1 expression and protein synthesis. Our data suggest that IL-13 regulates MMP-1 expression in response to TNF-α through an Akt-mediated pathway and may play a role in fibrotic diseases such as scleroderma.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid