Merck
Activation of cGAS/STING pathway upon paramyxovirus infection.
iScience (2021-06-19)
Mathieu Iampietro, Claire Dumont, Cyrille Mathieu, Julia Spanier, Jonathan Robert, Aude Charpenay, Sébastien Dupichaud, Kévin P Dhondt, Noémie Aurine, Rodolphe Pelissier, Marion Ferren, Stéphane Mély, Denis Gerlier, Ulrich Kalinke, Branka Horvat
ABSTRACT
During inflammatory diseases, cancer, and infection, the cGAS/STING pathway is known to recognize foreign or self-DNA in the cytosol and activate an innate immune response. Here, we report that negative-strand RNA paramyxoviruses, Nipah virus (NiV), and measles virus (MeV), can also trigger the cGAS/STING axis. Although mice deficient for MyD88, TRIF, and MAVS still moderately control NiV infection when compared with wild-type mice, additional STING deficiency resulted in 100% lethality, suggesting synergistic roles of these pathways in host protection. Moreover, deletion of cGAS or STING resulted in decreased type I interferon production with enhanced paramyxoviral infection in both human and murine cells. Finally, the phosphorylation and ubiquitination of STING, observed during viral infections, confirmed the activation of cGAS/STING pathway by NiV and MeV. Our data suggest that cGAS/STING activation is critical in controlling paramyxovirus infection and possibly represents attractive targets to develop countermeasures against severe disease induced by these pathogens.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%