Merck
  • Synthesis and pharmacological investigation of novel 4-(2-methylphenyl)-1-substituted-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-ones as new class of H(1)-antihistaminic agents.
Synthesis and pharmacological investigation of novel 4-(2-methylphenyl)-1-substituted-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-ones as new class of H(1)-antihistaminic agents.
European journal of medicinal chemistry (2007-11-23)
V Alagarsamy, M Rupeshkumar, K Kavitha, S Meena, D Shankar, A A Siddiqui, R Rajesh
ABSTRACT
A series of novel 1-substituted-4-(2-methylphenyl)-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-ones were synthesized by the cyclization of 2-hydrazino-3-(2-methylphenyl)-3H-quinazolin-4-one with various one carbon donors. The starting material 2-hydrazino-3-(2-methylphenyl)-3H-quinazolin-4-one was synthesized from 2-methyl aniline by a novel innovative route. The title compounds were tested for their in vivo H(1)-antihistaminic activity on guinea pigs; all the tested compounds protected the animals from histamine-induced bronchospasm significantly. Compound 1-methyl-4-(2-methylphenyl)-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-one (II) emerged as the most active compound of the series and it is more potent (72.45%) when compared to the reference standard chlorpheniramine maleate (71%). Compound II showed negligible sedation (11%) when compared to chlorpheniramine maleate (30%). Hence it could serve as the prototype molecule for further development as a new class of H(1)-antihistaminic agents.
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)