Merck
  • Effect of the interference with DRP1 expression on the biological characteristics of glioma stem cells.
Effect of the interference with DRP1 expression on the biological characteristics of glioma stem cells.
Experimental and therapeutic medicine (2021-05-15)
Linna Zhang, Huimei Cao, Hong Tao, Jijuan Yang, Wei Gong, Qikuan Hu
ABSTRACT
In the present study, a model of glioma stem cells (GSCs) was established and combined with molecular targeting drugs in order to observe its inhibitory effect on the proliferation and biological characteristics of GSCs, with the aim of providing a potential target for the treatment of glioma. On the basis of a relatively classical induction strategy with neuron induction medium, a large number of GSC-like cells in good condition and globular growth were amplified in vitro, which had the potential to differentiate into neurons, oligodendrocytes and astrocytes/glioma cells. It was observed that the interference with dynamin-related protein 1 expression using Mdivi-1, a mitochondrial mitotic inhibitor, at the optimal concentration, decreased the expression level of stem cell-associated genes, inhibited proliferation and promoted apoptosis in GSCs. The present study provided an experimental basis for a novel strategy of cancer treatment with tumor stem cells as the target.
Sigma-Aldrich
Monoclonal Anti-Oligodendrocyte Marker O4 antibody produced in mouse, clone O4, purified immunoglobulin, lyophilized powder