Merck
  • Analysis of the Pseudouridimycin Biosynthetic Pathway Provides Insights into the Formation of C-nucleoside Antibiotics.
Analysis of the Pseudouridimycin Biosynthetic Pathway Provides Insights into the Formation of C-nucleoside Antibiotics.
Cell chemical biology (2018-03-20)
Margherita Sosio, Eleonora Gaspari, Marianna Iorio, Silvia Pessina, Marnix H Medema, Alice Bernasconi, Matteo Simone, Sonia I Maffioli, Richard H Ebright, Stefano Donadio
ABSTRACT
Pseudouridimycin (PUM) is a selective nucleoside-analog inhibitor of bacterial RNA polymerase with activity against Gram-positive and Gram-negative bacteria. PUM, produced by Streptomyces sp. ID38640, consists of a formamidinylated, N-hydroxylated Gly-Gln dipeptide conjugated to 5'-aminopseudouridine. We report the characterization of the PUM gene cluster. Bioinformatic analysis and mutational knockouts of pum genes with analysis of accumulated intermediates, define the PUM biosynthetic pathway. The work provides the first biosynthetic pathway of a C-nucleoside antibiotic and reveals three unexpected features: production of free pseudouridine by the dedicated pseudouridine synthase, PumJ; nucleoside activation by specialized oxidoreductases and aminotransferases; and peptide-bond formation by amide ligases. A central role in the PUM biosynthetic pathway is played by the PumJ, which represents a divergent branch within the TruD family of pseudouridine synthases. PumJ-like sequences are associated with diverse gene clusters likely to govern the biosynthesis of different classes of C-nucleoside antibiotics.
Sigma-Aldrich
GenElute Gel Extraction Kit, sufficient for 70 purifications
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 70 purifications
Sigma-Aldrich
Thiostrepton from Streptomyces azureus, ≥90% (HPLC)