Merck
  • Profiling Protein Arginine Deiminase 4 (PAD4): a novel screen to identify PAD4 inhibitors.
Profiling Protein Arginine Deiminase 4 (PAD4): a novel screen to identify PAD4 inhibitors.
Bioorganic & medicinal chemistry (2007-10-30)
Bryan Knuckley, Yuan Luo, Paul R Thompson
ABSTRACT
Protein Arginine Deiminase 4 (PAD4) has emerged as a leading target for the development of a Rheumatoid Arthritis (RA) pharmaceutical. Herein, we describe the development of a novel screen for PAD4 inhibitors that is based on a PAD4-targeted Activity-Based Protein Profiling reagent, denoted Rhodamine-conjugated F-Amidine (RFA). This screen was validated by screening 10 Disease Modifying Anti-Rheumatic Drugs (DMARDs) and identified streptomycin, minocycline, and chlortetracycline as micromolar inhibitors of PAD4 activity.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Sulfapyridine, ≥99.0%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Supelco
Sulfamethoxazole
Supelco
Sulfapyridine, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Sigma-Aldrich
5-Aminosalicylic acid, 95%