Merck
  • Differential Signaling and Virus Production in Calu-3 Cells and Vero Cells upon SARS-CoV-2 Infection.
Differential Signaling and Virus Production in Calu-3 Cells and Vero Cells upon SARS-CoV-2 Infection.
Biomolecules & therapeutics (2021-01-29)
Byoung Kwon Park, Dongbum Kim, Sangkyu Park, Sony Maharjan, Jinsoo Kim, Jun-Kyu Choi, Madhav Akauliya, Younghee Lee, Hyung-Joo Kwon
ABSTRACT
Severe acute respiratory syndrome CoV-2 (SARS-CoV-2) is responsible for the current coronavirus disease 2019 (COVID-19) pandemic. Signaling pathways that are essential for virus production have potential as therapeutic targets against COVID-19. In this study, we investigated cellular responses in two cell lines, Vero and Calu-3, upon SARS-CoV-2 infection and evaluated the effects of pathway-specific inhibitors on virus production. SARS-CoV-2 infection induced dephosphorylation of STAT1 and STAT3, high virus production, and apoptosis in Vero cells. However, in Calu-3 cells, SARS-CoV-2 infection induced long-lasting phosphorylation of STAT1 and STAT3, low virus production, and no prominent apoptosis. Inhibitors that target STAT3 phosphorylation and dimerization reduced SARS-CoV-2 production in Calu-3 cells, but not in Vero cells. These results suggest a necessity to evaluate cellular consequences upon SARS-CoV-2 infection using various model cell lines to find out more appropriate cells recapitulating relevant responses to SARS-CoV-2 infection in vitro.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Tyrphostin AG 490, solid
Sigma-Aldrich
S3I-201, ≥97% (HPLC)
Sigma-Aldrich
PD 169316, ≥98% (HPLC), solid
Sigma-Aldrich
STA-21, ≥95% (HPLC)