Merck
  • Serine proteinase inhibition by the active site titrant N alpha-(N, N-dimethylcarbamoyl)-alpha-azaornithine p-nitrophenyl ester. A comparative study.
Serine proteinase inhibition by the active site titrant N alpha-(N, N-dimethylcarbamoyl)-alpha-azaornithine p-nitrophenyl ester. A comparative study.
European journal of biochemistry (2000-02-15)
P Ascenzi, G Balliano, C Gallina, F Polticelli, M Bolognesi
ABSTRACT
Kinetics for the hydrolysis of the chromogenic active-site titrant N alpha-(N,N-dimethylcarbamoyl)-alpha-azaornithine p-nitrophenyl ester (Dmc-azaOrn-ONp) catalysed by bovine beta-trypsin, bovine alpha-thrombin, bovine Factor Xa, human alpha-thrombin, human Factor Xa, human Lys77-plasmin, human urinary kallikrein, Mr 33 000 and Mr 54 000 species of human urokinase, porcine pancreatic beta-kallikrein-A and -B and Ancrod (the coagulating serine proteinase from the Malayan pit viper Agkistrodon rhodostoma venom) have been obtained between pH 6.0 and 8.0, at 21.0 degrees C, and analysed in parallel with those for the enzymatic cleavage of N alpha-(N,N-dimethylcarbamoyl)-alpha-azalysine p-nitrophenyl ester (Dmc-azaLys-ONp). The enzyme kinetics are consistent with the minimum three-step catalytic mechanism of serine proteinases, the rate-limiting step being represented by the deacylation process. Bovine beta-trypsin kinetics are modulated by the acid-base equilibrium of the His57 catalytic residue (pKa approximately 6.9). Dmc-azaOrn-ONp and Dmc-azaLys-ONp bind stoichiometrically to the serine proteinase active site, and allow the reliable determination of the active enzyme concentration between 1.0 x 10-6 M and 3.0 x 10-4 M. The affinity and the reactivity for Dmc-azaOrn-ONp (expressed by Ks and k+2/Ks, respectively) of the serine proteinases considered are much lower than those for Dmc-azaLys-ONp. The very different affinity and reactivity properties for Dmc-azaOrn-ONp and Dmc-azaLys-ONp have been related to the different size of the ornithine/lysine side chains, and to the ensuing different positioning of the active-site titrants upon binding to the enzyme catalytic centre (i.e. to P1-S1 recognition). These data represent the first detailed comparative investigation on the catalytic properties of serine proteinases towards an ornithine derivative (i. e. Dmc-azaOrn-ONp).
Sigma-Aldrich
Z-L-Lys-ONp hydrochloride
Pricing and availability is not currently available.