Merck
  • An ARF GTPase module promoting invasion and metastasis through regulating phosphoinositide metabolism.
An ARF GTPase module promoting invasion and metastasis through regulating phosphoinositide metabolism.
Nature communications (2021-03-14)
Marisa Nacke, Emma Sandilands, Konstantina Nikolatou, Álvaro Román-Fernández, Susan Mason, Rachana Patel, Sergio Lilla, Tamas Yelland, Laura C A Galbraith, Eva C Freckmann, Lynn McGarry, Jennifer P Morton, Emma Shanks, Hing Y Leung, Elke Markert, Shehab Ismail, Sara Zanivan, Karen Blyth, David M Bryant
ABSTRACT
The signalling pathways underpinning cell growth and invasion use overlapping components, yet how mutually exclusive cellular responses occur is unclear. Here, we report development of 3-Dimensional culture analyses to separately quantify growth and invasion. We identify that alternate variants of IQSEC1, an ARF GTPase Exchange Factor, act as switches to promote invasion over growth by controlling phosphoinositide metabolism. All IQSEC1 variants activate ARF5- and ARF6-dependent PIP5-kinase to promote PI(3,4,5)P3-AKT signalling and growth. In contrast, select pro-invasive IQSEC1 variants promote PI(3,4,5)P3 production to form invasion-driving protrusions. Inhibition of IQSEC1 attenuates invasion in vitro and metastasis in vivo. Induction of pro-invasive IQSEC1 variants and elevated IQSEC1 expression occurs in a number of tumour types and is associated with higher-grade metastatic cancer, activation of PI(3,4,5)P3 signalling, and predicts long-term poor outcome across multiple cancers. IQSEC1-regulated phosphoinositide metabolism therefore is a switch to induce invasion over growth in response to the same external signal. Targeting IQSEC1 as the central regulator of this switch may represent a therapeutic vulnerability to stop metastasis.
Sigma-Aldrich
Anti-ARFGAP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-PPP6C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-ARF6 antibody, Mouse monoclonal, clone ARFAG, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-LRP1 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.