Merck
NUPR1 is a critical repressor of ferroptosis.
Nature communications (2021-01-30)
Jiao Liu, Xinxin Song, Feimei Kuang, Qiuhong Zhang, Yangchun Xie, Rui Kang, Guido Kroemer, Daolin Tang
ABSTRACT
Ferroptosis is a type of iron-dependent regulated cell death, representing an emerging disease-modulatory mechanism. Transcription factors play multiple roles in ferroptosis, although the key regulator for ferroptosis in iron metabolism remains elusive. Using NanoString technology, we identify NUPR1, a stress-inducible transcription factor, as a driver of ferroptosis resistance. Mechanistically, NUPR1-mediated LCN2 expression blocks ferroptotic cell death through diminishing iron accumulation and subsequent oxidative damage. Consequently, LCN2 depletion mimics NUPR1 deficiency with respect to ferroptosis induction, whereas transfection-enforced re-expression of LCN2 restores resistance to ferroptosis in NUPR1-deficient cells. Pharmacological or genetic blockade of the NUPR1-LCN2 pathway (using NUPR1 shRNA, LCN2 shRNA, pancreas-specific Lcn2 conditional knockout mice, or the small molecule ZZW-115) increases the activity of the ferroptosis inducer erastin and worsens pancreatitis, in suitable mouse models. These findings suggest a link between NUPR1-regulated iron metabolism and ferroptosis susceptibility.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Anti-NUPR1 (C-terminal) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
ZZW-115 trihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
L-Arginine monohydrochloride, reagent grade, ≥98% (HPLC), powder