Merck
  • The ameliorative effect of sophoricoside on mast cell-mediated allergic inflammation in vivo and in vitro.
The ameliorative effect of sophoricoside on mast cell-mediated allergic inflammation in vivo and in vitro.
Molecules (Basel, Switzerland) (2013-05-24)
Su-Jin Kim, Gil-Yong Lee, Ji-Wook Jung, Sa-Rang Oh, Eun-Mi Ahn, Sung-Hoon Kim, Seung-Heon Hong, Jae-Young Um
ABSTRACT
Sophoricoside exhibits numerous pharmacological effects, including anti- inflammatory and anti-cancer actions, yet the exact mechanism that accounts for the anti-allergic effects of sophoricoside is not completely understood. The aim of the present study was to elucidate whether and how sophoricoside modulates the mast cell-mediated allergic inflammation in vitro and in vivo. We investigated the pharmacological effects of sophoricoside on both compound 48/80 or histamine-induced scratching behaviors and 2,4-dinitrochlorobenzene (DNCB)-induced atopic dermatitis in mice. Additionally, to find a possible explanation for the anti-inflammatory effects of sophoricoside, we evaluated the effects of sophoricoside on the production of histamine and inflammatory cytokines and activation of nuclear factor-κB (NF-κB) and caspase-1 in phorbol 12-myristate 13-acetate plus calcium ionophore A23187 (PMACI)-stimulated human mast cells (HMC-1). The finding of this study demonstrated that sophoricoside reduced compound 48/80 or histamine-induced scratching behaviors and DNCB-induced atopic dermatitis in mice. Additionally, sophoricoside inhibited the production of inflammatory cytokines as well as the activation of NF-κB and caspase-1 in stimulated HMC-1. Collectively, the findings of this study provide us with novel insights into the pharmacological actions of sophoricoside as a potential molecule for use in the treatment of allergic inflammation diseases.
Sigma-Aldrich
Sophoricoside, ≥98% (HPLC)
Pricing and availability is not currently available.