Merck
Role of Clathrin Light Chains in Regulating Invadopodia Formation.
Cells (2021-03-07)
Markus Mukenhirn, Francesco Muraca, Delia Bucher, Edgar Asberger, Elisa Cappio Barazzone, Elisabetta Ada Cavalcanti-Adam, Steeve Boulant
ABSTRACT
One of the most fundamental processes of the cell is the uptake of molecules from the surrounding environment. Clathrin-mediated endocytosis (CME) is the best-described uptake pathway and regulates nutrient uptake, protein and lipid turnover at the plasma membrane (PM), cell signaling, cell motility and cell polarity. The main protein in CME is clathrin, which assembles as a triskelion-looking building block made of three clathrin heavy chains and three clathrin light chains. Compared to clathrin heavy chains (CHCs), the role of the two isoforms of clathrin light chains (CLCA and CLCB) is poorly understood. Here, we confirm that the simultaneous deletion of both CLCA/B causes abnormal actin structures at the ventral PM and we describe them, for the first time, as functional invadopodia rather than disorganized actin-cytoskeleton assembly sites. Their identification is based on the occurrence of common invadopodia markers as well as functional invadopodia activity characterized by an increased local proteolytic activity of the extracellular matrix proteins. We demonstrate that CLCA/B deletion impacts the intracellular trafficking and recovery of the matrix metalloproteinase 14 (MMP14) leading to its accumulation at the plasma membrane and induction of invadopodia formation. Importantly, we show that invadopodia formation can be prevented by depletion of MMP14. As such, we propose that CLCA/B regulate invadopodia formation by regulating MMP14 delivery to the plasma membrane.
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Anti-Huntingtin-Interacting Protein 1 Related (HIP1R) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid