Merck
  • Increased concentration of serum periostin is associated with poor outcome of patients with aneurysmal subarachnoid hemorrhage.
Increased concentration of serum periostin is associated with poor outcome of patients with aneurysmal subarachnoid hemorrhage.
Journal of clinical laboratory analysis (2018-03-03)
Weijian Luo, Hao Wang, Jiliang Hu
ABSTRACT
To explore the role of serum periostin in patients with aneurysmal subarachnoid hemorrhage (aSAH). We conducted a retrospective study and 124 aSAH patients treated in Shenzhen People's hospital during March 1st 2015 to December 30th 2016 were included. Baseline information, neurological status and clinical outcome were recorded. Blood samples on admission were collected and enzyme linked immunosorbent assay (ELISA) kits were used to detect the serum level of periostin. Spearman's Correlation Analysis was used to analyze the correlation between periostin and clinical severity. Receiver operating characteristic (ROC) curve was performed to investigate variables' prognostic value in patients with aSAH. The average age of patients included was 57.23 years old. Preliminary analysis revealed that serum periostin was significantly correlated with clinical severity. Patients with poor outcome at 12 months had higher level of periostin than patients with good outcome. Multivariate logistic regression analysis showed elevated level of periostin was significantly associated with poor outcome and the AUC was 0.85 for periostin in predicting poor outcome of patient with aSAH. Elevated serum periostin concentrations are significantly associated with clinical severity and poor outcome of aSAH patients, which indicate serum periostin can be used as a prognostic biomarker in patients with aSAH.
Sigma-Aldrich
Human Periostin ELISA Kit, for cell culture supernatants, plasma, and serum samples