Merck
  • Extracellular vesicles are associated with C-reactive protein in sepsis.
Extracellular vesicles are associated with C-reactive protein in sepsis.
Scientific reports (2021-03-28)
Birgit Fendl, René Weiss, Tanja Eichhorn, Ingrid Linsberger, Taras Afonyushkin, Florian Puhm, Christoph J Binder, Michael B Fischer, Viktoria Weber
ABSTRACT
There is increasing evidence that C-reactive protein (CRP) can mediate inflammatory reactions following the transformation of functionally inert pentameric CRP (pCRP) into its structural isoform pCRP* and into monomeric CRP (mCRP). This conversion can occur on the membranes of apoptotic or activated cells or on extracellular vesicles (EVs) shed from the cell surface. Here, we characterized the association of CRP with EVs in plasma from sepsis patients using flow cytometry, and found highly elevated levels of total EV counts and CRP+ EVs as compared to healthy individuals. We further assessed the ability of PentraSorb CRP, an extracorporeal device for the adsorption of CRP, to deplete free CRP and CRP+ EVs. Treatment of septic plasma with the adsorbent in vitro resulted in almost complete removal of both, free CRP and CRP+ EVs, while total EV counts remained largely unaffected, indicating the detachment of CRP from the EV surface. EVs from septic plasma elicited a release of interleukin-8 from cultured human monocytes, which was significantly reduced by adsorbent treatment prior to EV isolation. Our findings provide evidence that CRP+ EVs exhibit pro-inflammatory characteristics and can contribute to the spreading of inflammation throughout the circulation on top of their pro-coagulant activity.
Sigma-Aldrich
Monoclonal Anti-C-Reactive Protein antibody produced in mouse, clone CRP-8, ascites fluid