Merck
  • Synthesis and biological activity of a 3,4,5-trimethoxybenzoyl ester analogue of epicatechin-3-gallate.
Synthesis and biological activity of a 3,4,5-trimethoxybenzoyl ester analogue of epicatechin-3-gallate.
Journal of medicinal chemistry (2008-03-08)
Luís Sánchez-del-Campo, Francisco Otón, Alberto Tárraga, Juan Cabezas-Herrera, Soledad Chazarra, José Neptuno Rodríguez-López
ABSTRACT
Despite presenting bioavailability problems, tea catechins have emerged as promising chemopreventive agents because of their observed efficacy in various animal models. To improve the stability and cellular absorption of tea polyphenols, we developed a new catechin-derived compound, 3- O-(3,4,5-trimethoxybenzoyl)-(-)-epicatechin (TMECG), which has shown significant antiproliferative activity against several cancer cell lines, especially melanoma. The presence of methoxy groups in its ester-bound gallyl moiety drastically decreased its antioxidant and prooxidant properties without affecting its cell-antiproliferative effects, and the data indicated that the 3-gallyl moiety was essential for its biological activity. As regards its action mechanism, we demonstrated that TMECG binds efficiently to human dihydrofolate reductase and down-regulates folate cycle gene expression in melanoma cells. Disruption of the folate cycle by TMECG is a plausible explanation for its observed biological effects and suggests that, like other antifolate compounds, TMECG could be of clinical value in cancer therapy.
Product Description
Supelco
(−)-Epicatechin, analytical standard
Supelco
(−)-Epigallocatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g