Merck
  • Simple Derivatization-Gas Chromatography-Mass Spectrometry for Fatty Acids Profiling in Soil Dissolved Organic Matter.
Simple Derivatization-Gas Chromatography-Mass Spectrometry for Fatty Acids Profiling in Soil Dissolved Organic Matter.
Molecules (Basel, Switzerland) (2020-11-18)
Neil Yohan Musadji, Claude Geffroy-Rodier
ABSTRACT
Dissolved organic matter is an important component of the global carbon cycle that allows the distribution of carbon and nutrients. Therefore, analysis of soil dissolved organic matter helps us to better understand climate change impacts as it is the most dynamic and reactive fraction in terrestrial ecosystems. Its characterization at the molecular level is still challenging due to complex mixtures of hundreds of compounds at low concentration levels in percolating water. This work presents simple methods, such as thermochemolysis- or derivatization-gas chromatography, as an alternative for the analysis of fatty acids in dissolved organic matter without any purification step. The variables of the protocols were examined to optimize the processing conditions for the C9-C18 range. As a proof of concept, fatty acid distributions of soil percolating water samples from a long-term field experiment were successfully assessed. The variability of dissolved organic acid distributions was pronounced through depth profile and soil treatment but no major change in composition was observed. However, although the optimization was done from C9 to C18, detection within the C6-C32 fatty acids range was performed for all samples.
Product Description
Supelco
Volatile Free Acid Mix, certified reference material, bottle of 100 mL
Pricing and availability is not currently available.